missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11a, Mouse anti-Human, Clone: HI111, BV650, BD
Description
Host Species: Mouse
Clone: HI111
Isotype: IgG1, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Flow cytometry
Specifications
Specifications
| Antigen | CD11a |
| Classification | Monoclonal |
| Clone | HI111 |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 100 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Target Species | Baboon, Cynomolgus, Human, Rhesus Monkey |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?