missing translation for 'onlineSavingsMsg'
Learn More

CD11b/c Mouse anti-Rat, PE-Cy7, Clone: OX-42, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 562222

Product Code. 15801739

  • ONLINE EXCLUSIVE PRICE
    197.00 €
    196.75 € / Each
    Save 0.25 €
    0.13% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The OX-42 monoclonal antibody specifically binds to the CR3 complement (C3bi) receptor found on most monocytes, granulocytes, macrophages, dendritic cells, and microglia. It appears to recognize a common epitope shared by CD11b and CD11c (integrin αM and αX chains). OX-42 antibody inhibits C3bi binding activity.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD11b/c
Monoclonal
0.2mg/mL
Itgam/Integrin, alpha M, C3bi receptor, CR3; Itgad/Integrin, alpha D
Mouse
Affinity Purified
RUO
Rat
IgG2a κ
Flow Cytometry
OX-42
PE-Cyanine7
Aqueous buffered solution containing ≤0.09% sodium azide.
Resident peritoneal cells from (PVG.RT1[c] x PVG.RT1[u]) and (PVG.RT1[c] x PVG.RT1[a]) F1-hybrid rat
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.