missing translation for 'onlineSavingsMsg'
Learn More

anti-CD11c; Clone: B-ly6; BD

Product Code. 15803099
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15803099 25 Tests Each
15893089 100 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15803099 Supplier BD Biosciences Supplier No. 563788

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Host Species: Mouse
Clone: B-ly6
Isotype: IgG1, κ
Species Reactivity: Hu
Immunogen: Spleen Cells from Human with Hairy Cell Leukemia

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD11c
Classification Monoclonal
Clone B-ly6
Conjugate Brilliant Ultraviolet 395
Host Species Mouse
Immunogen Spleen Cells from Human with Hairy Cell Leukemia
Purification Method Affinity Purified
Quantity 25 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.