missing translation for 'onlineSavingsMsg'
Learn More

CD138 Rat anti-Mouse, APC, Clone: 281-2, BD

Rat Monoclonal Antibody

173.00 € - 446.19 €

Specifications

Antigen CD138
Clone 281-2
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15801259
View Documents
BD Biosciences
561705
25 μg
173.00 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15829538
View Documents
BD Biosciences
558626
0.1 mg
ONLINE EXCLUSIVE PRICE
465.00 €
446.19 € / Each
Save 18.81 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 281-2 antibody reacts with the core protein of CD138 (Syndecan-1), a cell-surface, integral membrane heparan sulfate- and chondroitin sulfate-containing proteoglycan that binds to interstitial extracellular matrix molecules. Syndecan-1 is predominantly expressed on epithelial cells, where its expression correlates with normal epithelial organization. It is also expressed on B lymphocytes at specific stages during their differentiation: precursor B cells in the bone marrow and antibody-secreting cells, including plasma cells, but not mature peripheral B cells. It is thus implicated in mediating B cell-matrix interactions. CD138 expression is also regulated during embryonic development, and the molecule shows a tissue- specific structural polymorphism resulting from different post-translational modifications. The 281-2 antibody may be used to detect the differently glycosylated forms, because it reacts with the core protein. Furthermore, the mAb detects the Syndecan-1 ectodomain which is cleaved from cell surfaces by a metalloproteinase.

Flow Cytometry

Specifications

Specifications

CD138
0.2mg/mL
Monoclonal
Rat
Mouse
NAMRU mouse mammary gland epithelial cell line NMuMG
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
281-2
Flow Cytometry
APC
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
IgG2a κ
Affinity Purified
SYND1; Syndecan-1; syn-1; Sdc1; Sstn; synstatin
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.