missing translation for 'onlineSavingsMsg'
Learn More

CD15 Mouse anti-Human, Unlabeled, Clone: HI98, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 555400

Product Code. 15838138

  • ONLINE EXCLUSIVE PRICE
    135.00 €
    129.47 € / Each
    Save 5.53 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The HI98 monoclonal antibody specifically reacts with 3-fucosyl-N-acetyllactosamine (3-FAL), a 220 kDa carbohydrate structure, also called X-hapten, SSEA-1, CD15 or Lewis X. This structure is found on a variety of cell surface glycolipids and glycoproteins. 3-FAL is expressed on >95% of granulocytes, including neutrophils and eosinophils, and to a varying degree on monocytes, but not on lymphocytes or basophils. CD15 plays a role in mediating phagocytosis, bactericidal activity and chemotaxis. This antibody is also suitable for staining formalin-fixed, paraffin-embedded tissue sections without pretreatment. Since the Abs are recognizing a carbohydrate epitope (3-fucosyl-N-acetyllactosamine) they also should work across species and not only for human.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD15
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Human Mononuclear Cells
0.1 mg
Immunology
Human
IgM κ
Flow Cytometry
HI98
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.