missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-CD16; Clone: 3G8; BD
Description
Host Species: Mouse
Clone: 3G8
Isotype: IgG1, κ
Species Reactivity: Bb, Cyn, Hu, Rm
Immunogen: Human polymorphonuclear leukocytes
Flow cytometry
Specifications
Specifications
| Antigen | CD16 |
| Classification | Monoclonal |
| Clone | 3G8 |
| Conjugate | Brilliant Ultraviolet 395 |
| Host Species | Mouse |
| Immunogen | Human polymorphonuclear leukocytes |
| Purification Method | Affinity Purified |
| Quantity | 25 Tests |
| Regulatory Status | RUO |
| Primary or Secondary | Primary |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?