Learn More
CD19 Mouse anti-Human, Alexa Fluor 700, Clone: HIB19, BD
Description
Reacts with the 95kDa type I transmembrane glycoprotein expressed during all stages of B-cell differentiation and maturation, except on plasma cells. CD19 is also present on follicular dendritic cells. It is not found on T cells or on normal granulocytes. CD19 plays a role in regulation of B-cell proliferation. It associates with the complement receptor 2 (CD21), TAPA-1 (CD81), Leu 13, and/or MHC class II to form a signal transduction complex on the surface of B cells. Anti-CD19 clone HIB19 partially blocks clone B43, another CD19 mAb. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Mouse
Clone: HIB19
Isotype: IgG1, κ
Cross Reactivity: Human
Flow Cytometry
Specifications
Specifications
| Antigen | CD19 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | HIB19 |
| Concentration | 0.2mg/mL |
| Conjugate | Alexa Fluor 700 |
| Formulation | Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. |
| Host Species | Mouse |
| Purification Method | Affinity Purified |
| Quantity | 25 μg |
| Show More |
For Research Use Only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.