missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD253 Mouse anti-Human, Unlabeled, Clone: RIK-2, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 550515
This item is not returnable.
View return policy
Description
The RIK-2 monoclonal antibody specifically binds to CD253 which is also known as TRAIL (TNF-Related Apoptosis-Inducing Ligand) and Apo2 ligand (APO-2L). CD253 is a member of the Tumor Necrosis Factor Superfamily and is encoded by the TNFSF10 gene. CD253 is a type II membrane protein that may be expressed as a full-length, cell surface associated protein as well as in a soluble form. Both surface and soluble forms of TRAIL rapidly induce apoptosis by a wide range of cell lines but not by normal cells. TRAIL is expressed by activated T cells, NK cells, dendritic cells, monocytes and by a variety of non-lymphoid cells. TRAIL can bind to and exert apoptosis through DR4 (TRAIL-R1) and DR5 (TRAIL-R2) receptors. It can also bind to decoy receptors, including DcR1/TRID/TRAIL-R3 and DcR2/TRUNDD/TRAIL-R4, and possibly OPG/TNFRSF11B, which may serve to regulate TRAIL activity. TRAIL has been shown to be involved in T cell-mediated cytotoxicity, however its mechanism of action remains to be fully elucidated. The RIK-2 clone was selected based on its ability to block TRAIL-mediated cytotoxic activity.
Specifications
| CD253 | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Human TRAIL | |
| 0.1 mg | |
| Immunology | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| RIK-2 | |
| Unconjugated | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only.
Spot an opportunity for improvement?Share a Content Correction