missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CD4 PerCP-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
200.00 € - 331.00 €
Specifications
| Antigen | CD4 |
|---|---|
| Clone | 11830 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18771749
![]() |
R&D Systems
FAB3791C-025 |
25 Tests |
200.00 €
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18779806
![]() |
R&D Systems
FAB3791C-100 |
100 Tests |
331.00 €
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD4 Monoclonal specifically detects CD4 in Human samples. It is validated for Flow Cytometry.Specifications
| CD4 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD4, CD4 antigen, CD4 antigen (p55), CD4 molecule, CD4 receptor, CD4mut, T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4 | |
| CD4 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human CD4 in direct ELISAs. |
| 11830 | |
| Flow Cytometry | |
| PerCP | |
| Mouse | |
| Human | |
| P01730 | |
| 920 | |
| Recombinant Human CD4. Extracellular Domain Accession # P01730 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title