Learn More
CD44 Mouse anti-Human, APC, Clone: G44-26 (also known as C26), BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 560890
Description
The G44-26 monoclonal antibody specifically binds to the 80-95 kDa, glycosylated type I transmembrane protein, also known as phagocytic glycoprotein-1 (Pgp-1). CD44 is the receptor for hyaluronic acid. CD44 is expressed on leucocytes, erythrocytes, epithelial cells and weakly on platelets. CD44 is also called extracellular matrix receptor type III and has functional roles in cell migration, lymphocyte homing and adhesion during hematopoiesis and lymphocyte activation. This antibody is suitable for staining formalin-fixed, paraffin-embedded tissue sections using citrate buffer pretreatment. This antibody recognizes the epitope 1 of CD44 antigen according to the HLDA workshop studies.
Specifications
| CD44 | |
| Monoclonal | |
| APC | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG2b κ |
| Flow Cytometry | |
| G44-26 (also known as C26) | |
| Phagocytic glycoprotein 1; Pgp-1; H-CAM; Hermes; ECMR III; HUTCH-1 | |
| Mouse | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.