missing translation for 'onlineSavingsMsg'
Learn More

anti-CD44, BV711; Clone: IM7; BD

Product Code. 15843249
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15843249 50 μg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15843249 Supplier BD Biosciences Supplier No. 563971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Host Species: Rat
Clone: IM7
Isotype: IgG1, κ
Species Reactivity: Ms
Immunogen: Dexamethasone-induced cells of the SJL Mouse spontaneous myeloid leukemia M1

Flow cytometry

TRUSTED_SUSTAINABILITY

Specifications

Antigen CD44
Classification Monoclonal
Clone IM7
Concentration 0.2mg/mL
Host Species Rat
Immunogen Dexamethasone-induced cells of the SJL Mouse spontaneous myeloid leukemia M1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.