missing translation for 'onlineSavingsMsg'
Learn More

CD71 Mouse anti-Human, Unlabeled, Clone: M-A712, BD

Product Code. 15838248 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15838248 0.1 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15838248 Supplier BD Biosciences Supplier No. 555534
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Mouse Monoclonal Antibody

The M-A712 monoclonal antibody specifically binds to CD71 which is also known as the transferrin receptor (TFR). This type II transmembrane glycoprotein is expressed on cells as a disulfide-linked homodimer comprised of 95 kDa monomers. CD71 is expressed on activated lymphocytes, monocytes, macrophages, erythroid progenitors, brain endothelium, and most proliferating cells. CD71 is not expressed on resting lymphocytes and is upregulated during lymphocyte responses to antigens or mitogens. Through an endocytic pathway, the transferrin receptor mediates cellular iron uptake by binding and internalizing iron that is bound to transferrin. After releasing iron within the low pH endosomal environment, transferrin and its receptor can be recycled to the cell surface.

TRUSTED_SUSTAINABILITY

Tekniske data

Antigen CD71
Applications Flow Cytometry
Classification Monoclonal
Clone M-A712
Concentration 0.5mg/mL
Conjugate Unconjugated
Description TFR; TFRC; Trfr; TfR1; Transferrin receptor protein 1
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen T-CLL and Jurkat Cells
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Vis mere Vis mindre

For Research Use Only.

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.