missing translation for 'onlineSavingsMsg'
Learn More

CDC27 Mouse, Unlabeled, Clone: 35, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610454

 View more versions of this product

Product Code. 15865469

  • ONLINE EXCLUSIVE PRICE
    226.00 €
    216.14 € / 1 set
    Save 9.86 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CDC16 and CDC27 were discovered in yeast as genes essential for the G2/M transition of the cell cycle. The human homologues of CDC16 and CDC27were cloned and found to encode proteins of 72kDa and 97kDa, respectively. Biochemical and subcellular localization experiments have shown that CDC16 and CDC27 are novel components of the mitotic spindle centrosome in human cells. In HeLa cells, there are about 100,000 copies of each protein per cell. Although its precise role during mitosis remains to be determined, CDC16 appears to be required for cyclin degradation in yeast. Wild type CDC27 is required for the initiation of anaphase. Immunofluorescence studies have shown that both proteins are components of the centrosome in the absence of spindle fibers.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

CDC27
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Human, Mouse, Rat
IgG2b
Western Blot
35
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.