missing translation for 'onlineSavingsMsg'
Learn More

anti-CENPP, Matched Antibody Pair, Abnova

Product Code. 16143934
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16143934 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16143934 Supplier Abnova Supplier No. H00401541AP51.1Set
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Includes

Capture antibody: Mouse monoclonal anti-CENPP (100μg); Detection antibody: Rabbit purified polyclonal anti-CENPP (50μg); Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols

This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CENPP.

Host Species: Mouse, Rabbit
Species Reactivity: Human

ELISA

Specifications

Antigen CENPP
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Unconjugated
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human CENPP.
Gene CENPP
Gene Alias CENP-P/FLJ33928
Gene Symbols CENPP
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401541
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-CENPP (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-CENPP (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.