missing translation for 'onlineSavingsMsg'
Learn More

Chk2 Mouse, Unlabeled, Clone: 19, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 611570

 View more versions of this product

Product Code. 15855979

  • ONLINE EXCLUSIVE PRICE
    196.00 €
    187.80 € / 1 set
    Save 8.20 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

The cell cycle is regulated by multiple checkpoints that determine cell fate. Such checkpoints ensure that DNA replication and chromosomal segregation are completed with high fidelity. When DNA is damaged, specific kinases and phosphatases, key components of cell cycle checkpoints, function to arrest the cell cycle and provide the necessary time for DNA repair. For example, DNA damage induces arrest of the cell cycle at the G2 checkpoint via Wee1-mediated inhibitory phosphorylation of the kinase Cdc2. Inhibition is relieved by the Cdc25C
phosphatase via Cdc2 dephosphorylation. In turn, the Chk1 and Chk2 protein kinases phosphorylate and inhibit Cdc25C, thus preventing activation of the Cdc2-cyclin B complex and entry into mitosis. Chk2, the human homolog of S. cerevisiae Rad53 and S. pombe Cds1, contains a C-terminal kinase domain, an N-terminal regulatory region that is rich in TQ and SQ pairs, and a forked head-associated domain (FHA) which is found in other cell cycle kinases. Chk2 is expressed during late G1 to M phase and is found in the nucleoplasm. Thus, Chk1
and Chk2 activity may be an important checkpoint that inhibits entry into mitosis in response to DNA damage.

Host Species: Mouse
Clone: 19
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse Chk2 aa. 31-234
Formula Weight [Chemical]: 60kDa

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Chk2
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse Chk2 aa. 31-234
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Western Blot
19
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.