Learn More
CHST9 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530842
Code : A0
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, mouse brain. Predicted reactivity: Mouse (91%), Rat (90%), Rhesus Monkey (99%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Sulfate groups in carbohydrates confer highly specific functions on glycoproteins, glycolipids, and proteoglycans and are critical for cell-cell interaction, signal transduction, and embryonic development. Sulfotransferases, such as CHST9, carry out sulfation of carbohydrates (Hiraoka et al., 2001 ).Specifications
CHST9 | |
Polyclonal | |
Unconjugated | |
CHST9 | |
5430438D01Rik; carbohydrate (N-acetylgalactosamine 4-0) sulfotransferase 9; carbohydrate sulfotransferase 9; CHST9; GalNAc-4-O-sulfotransferase 2; GalNAc4ST2; galNAc-4-ST2; GalNAc4ST-2; GalNAc-4-sulfotransferase 2; N-acetylgalactosamine 4-O-sulfotransferase 2; N-acetylgalactosamine-4-O-sulfotransferase 2; UNQ2549/PRO6175 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
71367, 83539 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q76EC5, Q7L1S5 | |
CHST9 | |
Recombinant fragment corresponding to a region within amino acids 200 and 435 of Human CHST9 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |