Learn More
Cox-2 Mouse, Unlabeled, Clone: 33, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 610204
Description
Cyclooxygenase (Cox) is also known as prostaglandin H synthase or PGH synthase (E.C. 1.14.99.1). It catalyzes the conversion of arachidonate to prostaglandin H2 (PGH2), the precursor of PGE2, PGF2α, PGD2, prostacyclin, and thromboxane A2. Cox actually has two different enzymatic activities: a cyclooxygenase that mediates the formation of PGG2 from oxygen and arachidonate and a hydroperoxidase that catalyzes a reduction of PGG2 yielding PGH2. Two Cox genes, Cox-1 and Cox-2, have been isolated in several species. A 4kb mRNA encodes the 604 amino acid Cox-2 protein. The two human Cox isoenzymes are 61% identical in amino acid composition with the active sites being highly conserved. Cox-2 mRNA and protein levels are induced by serum, lipopolysaccharides, growth factors, human chorionic gonadotropin and phorbol testers in various mammalian cell types. It has been shown that interleukin-1α (IL-1α) induces increased levels of Cox-2 mRNA and protein in human endothelial cells. The sustained increase in Cox-2 is apparently due (at least in part) to IL-1α increasing the stability of Cox-2 mRNA. This type of regulatory mechanism may play an important role in chronic inflammatory conditions.
Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting
Specifications
| Cox-2 | |
| Monoclonal | |
| 250μg/mL | |
| PGHS-2, Cyclooxygenase-2 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at -20°C. |
| Western Blot | |
| 33 | |
| Unconjugated | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| 150 μg | |
| Cell Biology | |
| Chicken, Human, Mouse | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.