missing translation for 'onlineSavingsMsg'
Learn More

Crk Mouse, Unlabeled, Clone: 22, BD

Mouse Monoclonal Antibody

178.69 € - 196.45 €

Specifications

Antigen Crk
Clone 22
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15855219
View Documents
BD Biosciences
610035
50 μg
ONLINE EXCLUSIVE PRICE
187.00 €
178.69 € / 1 set
Save 8.31 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15865219
View Documents
BD Biosciences
610036
150 μg
ONLINE EXCLUSIVE PRICE
205.00 €
196.45 € / 1 set
Save 8.55 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Crk was first isolated as the v- crk oncogene from chicken retroviruses CT10 and ASV-1. All human cell lines examined to date express a 40 kDa Crk protein. In addition, there is variable expression of 42 kDa and 28 kDa Crk proteins. The c- crk gene is one of a class of genes, such as Nck and GRB2/ASH , which encode proteins that consist mainly of SH2 and SH3 domains. These proteins function as adaptor molecules in tyrosine kinase signal transduction pathways. The SH2 domains interact with phosphotyrosine-containing peptides, while the SH3 domains can enhance this interaction and/or bind to other cellular components. Both the SH2 and SH3 domains of the human Crk protein are required for differentiation of PC12 cells. Thus, Crk has a role in an NGF-induced signaling pathway that involves activation of p21ras. Furthermore, three proteins of 118 kDa, 125 kDa, and 136 kDa which specifically bind to the Crk SH3 domain have been identified.
Specifications

Specifications

Crk
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG2a
Affinity Purified
22
Western Blot
Unconjugated
RUO
Bovine, Canine, Chicken, Frog, Human, Mouse, Rat
Human Crk aa. 102 to 304
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.