missing translation for 'onlineSavingsMsg'
Learn More

CTCF Mouse, Unlabeled, Clone: 48, BD

Mouse Monoclonal Antibody

253.48 € - 476.00 €

Specifications

Antigen CTCF
Clone 48
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15886109
View Documents
BD Biosciences
612148
50 μg
ONLINE EXCLUSIVE PRICE
265.00 €
253.48 € / 1 set
Save 11.52 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15896109
View Documents
BD Biosciences
612149
150 μg
476.00 €
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The family of nucleic acid-binding C2H2 type zinc finger transcription factors is divided into two classes. One class consists of small proteins (Gli1, Krox-20, WT1, Egr-1, and Sp1) with conserved zinc finger clusters of 3 to 5 units, while the other class (ZNF91, ZNF74, ZFP37, CTCF) can contain more than 10 zinc finger clusters. CTCF is a ubiquitously expressed, highly conserved transcription factor that contains 10 C2H2- and 1 C2H-type zinc-finger motifs. CTCF binds to and represses transcription at the promoter-proximal regions of c-myc oncogenes,
while CTCF can bind to and activate transcription at the promoter for amyloid β. Another mechanism of CTCF-mediated repression may include binding to insulator regions between enhancers and promoters resulting in enhancer blocking. In addition, CTCF gene may be a candidate tumor suppressor gene, since it localizes to a narrow cancer-associated chromosome region, and has been shown to have tumor-specific rearrangements in breast cancer patients. Thus, CTCF transcriptional regulation may be important for cell cycle progression, differentiation, apoptosis, and tumorigenesis.

Immunofluorescence, Western Blotting

Specifications

Specifications

CTCF
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
48
Western Blot
Unconjugated
RUO
Human, Mouse, Rat
Human CTCF aa. 184-290
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.