missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human CXCL5/ENA-78 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
211.00 € - 2100.00 €
Specifications
| Antigen | CXCL5/ENA-78 |
|---|---|
| Clone | 33160 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18704438
![]() |
R&D Systems
MAB254-SP |
25 μg |
211.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18769448
![]() |
R&D Systems
MAB254-100 |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18724968
![]() |
R&D Systems
MAB254-500 |
500 μg |
2100.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CXCL5/ENA-78 Monoclonal specifically detects CXCL5/ENA-78 in Human samples. It is validated for Western Blot, Immunohistochemistry, Neutralization.Specifications
| CXCL5/ENA-78 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Neutralization 3-15 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P42830 | |
| 6374 | |
| E. coli-derived recombinant human CXCL5/ENA-78 Ala37-Asn114 (predicted) Accession # P42830 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human CXCL5/ENA-78 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) CXCL1, 2, 3, 8, 10, or recombinant mouse CXCL2 is observed. |
| 33160 | |
| Western Blot, Immunohistochemistry, Neutralization | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| chemokine (C-C motif) ligand 5, C-X-C motif chemokine 5, CXCL5, ENA78, ENA-78, ENA-78(1-78), Epithelial-derived neutrophil-activating protein 78, Neutrophil-activating peptide ENA-78, neutrophil-activating protein 78, SCYB5ENA78 | |
| CXCL5 | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title