missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DAZAP1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA541954
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: QAAPDMSKPP TAQPDFPYGQ YGLGSYSPAP PGCGPHFVYS LMVRLSSDVA Sequence homology: Human: 100%
DAZAP1 Polyclonal antibody specifically detects DAZAP1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot.Specifications
DAZAP1 | |
Polyclonal | |
Unconjugated | |
DAZAP1 | |
2410042M16Rik; DAZ associated protein 1; DAZAP1; DAZ-associated protein 1; Deleted in azoospermia-associated protein 1; LOC561459 protein; LOW QUALITY PROTEIN: DAZ-associated protein 1; MGC19907; mPrrp; testicular tissue protein Li 50 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
26528 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q96EP5 | |
DAZAP1 | |
Synthetic peptide directed towards the C-terminal of human DAZAP1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |