Learn More
DYKDDDDK Tag Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1984B
Code : LE
Additional Details : Weight : 1.24740kg
Description
PA1-984B detects the DYKDDDDK epitope tag sequence, similar to the FLAG tag from Sigma. PA1-984B has been successfully used in Western blot and immunofluorescence procedures to detect the presence of fusion proteins containing the epitope tag sequence DYKDDDDK. PA1-984B immunizing peptide corresponds to the epitope tag sequence DYKDDDDKC. This peptide (Cat. # PEP-087) is available for use in neutralization and control experiments.
DYKDDDDK Tag Polyclonal antibody specifically detects DYKDDDDK Tag in Tag samples. It is validated for Immunocytochemistry, Western Blot.Specifications
DYKDDDDK Tag | |
Polyclonal | |
Unconjugated | |
DDDDK; DDDDK epitope tag; ECS epitope tag; ECS tag; FLAG; FLAG Tag | |
Synthetic Peptide: D Y K D D D D K C | |
200 μg | |
Primary | |
Antibody | |
IgG |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Tag |