missing translation for 'onlineSavingsMsg'
Learn More

EB1 Mouse, Unlabeled, Clone: 5, BD

Mouse Monoclonal Antibody

186.18 € - 476.00 €

Specifications

Antigen EB1
Clone 5
Concentration 250μg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15815509
View Documents
BD Biosciences
610534
50 μg
ONLINE EXCLUSIVE PRICE
194.00 €
186.18 € / 1 set
Save 7.82 €
4% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15825509
View Documents
BD Biosciences
610535
150 μg
476.00 €
1 set
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Familial adenomatous polyposis is an autosomal inherited disease characterized by mutations in the APC (adenomatous polyposis coli) gene. Individuals with this disease are predisposed to colorectal polyps and cancer. It has been proposed that disruption of the APC gene plays a role in the disease. APC is a large coiled-coil protein that interacts, via its carboxy terminus, with catenins and with the protein, EB1. This 30kDa protein was discovered as a result of its strong binding to the C-terminal domain of APC. Like the APC-binding proteins β-Catenin and pp120Cas, EB1 may play an essential role in modulating APC activity and tumorigenesis.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

EB1
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1
Affinity Purified
5
Western Blot
Unconjugated
RUO
Chicken, Human, Mouse, Rat
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.