Learn More
anti-ECHS1 Matched Antibody Pair, Abnova
Description
Includes
Rabbit MaxPab™ affinity purified polyclonal anti-ECHS1 (100μg); Mouse monoclonal anti-ECHS1, IgG1κ (20μg); Sufficient for at least 3-5 x 96 well plates using recommended protocols.
The protein encoded by this gene functions in the second step of the mitochondrial fatty acid β-oxidation pathway. It catalyzes the hydration of 2-trans-enoyl-coenzyme A (CoA) intermediates to L-3-hydroxyacyl-CoAs. The gene product is a member of the hydratase/isomerase superfamily. It localizes to the mitochondrial matrix. Transcript variants utilizing alternative transcription initiation sites have been described in the literature.
ELISA
Specifications
Specifications
| Antigen | ECHS1 |
| Applications | ELISA |
| Classification | Monoclonal/Polyclonal |
| Conjugate | Unconjugated |
| Description | This antibody pair set comes with matched antibody pair to detect and quantify protein level of human ECHS1. |
| Gene | ECHS1 |
| Gene Alias | SCEH |
| Gene Symbols | ECHS1 |
| Host Species | Mouse, Rabbit |
| Purification Method | Affinity Purified |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.