missing translation for 'onlineSavingsMsg'
Learn More

EFP Mouse anti-Human, Unlabeled, Clone: 2, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610570

Product Code. 15825529

  • ONLINE EXCLUSIVE PRICE
    194.00 €
    186.18 € / 1 set
    Save 7.82 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Steriod hormones, unlike polypeptide hormones, diffuse inside the cell, bind cytoplasmic or nuclear receptors, and activate specific genes. Complete identification of such receptors would greatly increase the understanding of the celluar processes activated by steriods. EFP (Estrogen receptor Finger Protein) was identified as a result of its specific interaction with the estrogen receptor. EFP contains a zinc-finger domain, indicating that it is a DNA-binding protein. EFP transcription is regulated by estrogens and, in combination with the estrogen receptor, it may modulate the expression of estrogen-responsive genes.

This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

EFP
Monoclonal
250μg/mL
Estrogen receptor Finger Protein
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
2
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Human
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.