missing translation for 'onlineSavingsMsg'
Learn More

EphA4/Sek Mouse, Unlabeled, Clone: 35, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 610471

Product Code. 15885479

  • ONLINE EXCLUSIVE PRICE
    212.00 €
    203.30 € / 1 set
    Save 8.70 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

EphA4/Sek is a glycosylated protein tyrosine kinase of 986 amino acids and an apparent 110-130kDa molecular weight (molecular weight depends on glycocylation level of protein). EphA4 belongs to the Eph subfamily of receptor tyrosine kinases and like many others, it has an amino-terminal ligand-binding domain exposed to the extracellar matrix. EphA4 is predominantly expressed during development (this is common in the Eph family) and because of its mRNA localization, it has been proposed that it may play a role during segmentation of the hindbrain and mesoderm. Tyrosine 602 is a major autophosphorylation site of EphA4 and appears to be the docking site for the intracellular tyrosine kinase fyn.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

EphA4/Sek
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
35
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.