missing translation for 'onlineSavingsMsg'
Learn More

Fibronectin Mouse, Unlabeled, Clone: 10, BD

Mouse Monoclonal Antibody

172.06 € - 338.98 €

Specifications

Antigen Fibronectin
Clone 10
Concentration 250μg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15805249
View Documents
BD Biosciences
610077
50 μg
ONLINE EXCLUSIVE PRICE
179.00 €
172.06 € / 1 set
Save 6.94 €
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15815249
View Documents
BD Biosciences
610078
150 μg
ONLINE EXCLUSIVE PRICE
353.00 €
338.98 € / 1 set
Save 14.02 €
3% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The 240kDa dimeric fibronectin protein exists in two forms: a soluble protomer in body fluids and an insoluble multimer in the extracellular matrix. The latter is the primary functional form and creates a substrate for cell migration, a role which makes fibronectin vital to embryogenesis and wound response. Fibronectin mediates cytoskeletal organization, cell attachment, and cellular signaling through interactions with cellular receptors. Although various isoforms of fibronectin are derived by alternative splicing, they share a common N-terminus which is a critical region for cell surface binding in an initial step of multimer assembly. Further polymerization steps are regulated by fibronectin/integrin interactions and result in generation of the complex fibrils that constitute the fibronectin matrix.

Host Species: Mouse
Clone: 10
Isotype: IgG1
Species Reactivity: Human
Immunogen: Human Fibronectin
Formula Weight [Chemical]: 240kDa

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Specifications

Specifications

Fibronectin
250μg/mL
Monoclonal
Mouse
Cell Biology
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
IgG1 κ
Affinity Purified
FN; LETS
10
Flow Cytometry
Unconjugated
RUO
Bovine, Canine, Chicken, Human, Mouse, Rat
Human Fibronectin
Primary
Store undiluted at -20°C.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.