Learn More
anti-FKBP5 Matched Antibody Pair, Abnova
Brand: Abnova H00002289-AP11.1Set
Includes:
- 1. Capture antibody: rabbit affinity purified polyclonal anti-FKB5 (100μg)
- 2. Detection antibody: mouse monoclonal anti-FKBP5 κ (20μg)
- Sufficient for at least 3-5 x 96 well plates using recommended protocols.
Description
The protein encoded by this gene is a member of the immunophilin protein family, which play a role in immunoregulation and basic cellular processes involving protein folding and trafficking. This encoded protein is a cis-trans prolyl isomerase that binds to the immunosuppressants FK506 and rapamycin. It is thought to mediate calcineurin inhibition. It also interacts functionally with mature hetero-oligomeric progesterone receptor complexes along with the 90 kDa heat shock protein and P23 protein. This gene has been found to have multiple polyadenylation sites.
- Species Reactivity: Human
ELISA
Specifications
| FKBP5 | |
| Monoclonal/Polyclonal | |
| This antibody pair set comes with matched antibody pair to detect and quantify protein level of human FKBP5. | |
| FKBP51/FKBP54/MGC111006/P54/PPIase/Ptg-10 | |
| Mouse, Rabbit | |
| Affinity Purified | |
| RUO | |
| 2289 | |
| Human | |
| IgG1 |
| ELISA | |
| Unconjugated | |
| FKBP5 | |
| FKBP5 | |
| NA; FKBP5 (AAH42605, 1 a.a. ∽ 458 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 1 Set | |
| Primary | |
| Antibody pair set content: 1. Capture antibody: rabbit MaxPab™ affinity purified polyclonal anti-FKBP5 (100 μg) 2. Detection antibody: mouse monoclonal anti-FKBP5, IgG1 Kappa (20 μg) *Reagents are sufficient for at least 1-2 x 96 well plates using recommended protocols. | |
| Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.