missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FZD4 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Brand: Invitrogen PA518886
Code : A0
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 16,000.
FZD4 is a member of the frizzled gene family. Members of this family encode seven-transmembrane domain proteins that are receptors for the Wingless type MMTV integration site family of signaling proteins. Most frizzled receptors are coupled to the beta-catenin canonical signaling pathway. This protein may play a role as a positive regulator of the Wingless type MMTV integration site signaling pathway. A transcript variant retaining intronic sequence and encoding a shorter isoform has been described, however, its expression is not supported by other experimental evidence. Frizzled 4 is widely expressed, particularly in heart, skeletal muscle, ovary, and fetal kidney. Transcripts have also been identified in liver, kidney, pancreas, spleen, and fetal lung, and in smaller amounts in placenta, adult lung, prostate, testis, colon, and fetal brain.Specifications
FZD4 | |
Polyclonal | |
Unconjugated | |
FZD4 | |
CD344; CD344 antigen; EVR1; FEVR; frizzled 4, seven transmembrane spanning receptor; frizzled class receptor 4; frizzled family receptor 4; frizzled homolog 4; frizzled homolog 4 (Drosophila); frizzled receptor 4; frizzled-4; Fz4; Fz-4; Fzd4; FZD4S; FzE4; GPCR; hFz4; mFz4; MGC34390; rFz4; WNT receptor frizzled-4 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
8322 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q9ULV1 | |
FZD4 | |
Synthetic peptide sequence (DEEERRCDPIRIS) corresponding to the internal amino acids of FZD4 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |