missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GDF10 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531085
This item is not returnable.
View return policy
Description
Recommended positive controls: 293T, Raji, NIH-3T3. Predicted reactivity: Mouse (85%), Rat (85%), Bovine (89%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is a member of the bone morphogenetic protein (BMP) family and the TGF-beta superfamily. This group of proteins is characterized by a polybasic proteolytic processing site which is cleaved to produce a mature protein containing seven conserved cysteine residues. The members of this family are regulators of cell growth and differentiation in both embryonic and adult tissues. Studies in mice suggest that the protein encoded by this gene plays a role in skeletal morphogenesis.
Specifications
| GDF10 | |
| Polyclonal | |
| Unconjugated | |
| GDF10 | |
| BIP; Bmp3b; BMP-3B; Bone morphogenetic protein 3B; bone morphogenetic protein 3B; growth/differentiation factor 10; bone-inducing protein; Gdf10; GDF-10; growth differentiation factor 10; growth/differentiation factor 10; prepro bone inducing protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14560, 2662, 79216 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| P55107, P55108, P97737 | |
| GDF10 | |
| Recombinant protein encompassing a sequence within the center region of human GDF10. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction