Learn More
GFAP, Human, Matched Antibody Pair (AP52), Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00002670-AP52.1Set
Product Code. 16172094
  • 835.00 € / 1 set
Includes: Capture antibody: mouse monoclonal anti-GFAP, IgG2a Kappa (100μg) , Detection antibody: rabbit purified polyclonal anti-GFAP (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
GFAP
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human GFAP.
FLJ45472
Mouse, Rabbit
RUO
2670
Human
Liquid
ELISA
Unconjugated
GFAP
GFAP
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-GFAP, IgG2a Kappa (100 ug) 2. Detection antibody: rabbit purified polyclonal anti-GFAP (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.