Learn More
GFAP Mouse, PE, Clone: 1B4, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 561483
Description
GFAP (Glial Fibrillary Acidic Protein) is the major protein of glial filaments in differentiated astrocytes. BD Biosciences offers a panel of monoclonal antibodies (4A11, 1B4, 2E1) that specifically recognize GFAP. They do not cross-react with other intermediate filaments such as vimentin, neurofilament proteins, desmin, keratin, neurotubules or microfilaments.
Specifications
| GFAP | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Cow spinal cord homogenate | |
| 50 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Flow Cytometry | |
| 1B4 | |
| Glial Fibrillary Acidic Protein, FLJ45472 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Bovine, Canine, Chicken, Guinea Pig, Human, Mouse, Pig, Rabbit, Rat, Sheep | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.