missing translation for 'onlineSavingsMsg'
Learn More

Human gp130 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 3 publications

209.00 € - 1990.00 €

Specifications

Antigen gp130/CD130
Clone 29104
Dilution Western Blot 1 ug/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18751289
GREENER_CHOICE
View Documents
R&D Systems
MAB2281-SP
25 μg
209.00 €
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18708809
GREENER_CHOICE
View Documents
R&D Systems
MAB2281-100
100 μg
486.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18723328
GREENER_CHOICE
View Documents
R&D Systems
MAB2281-500
500 μg
1990.00 €
500µg
Ver productos alternativos
missing translation for 'toSaveUpto' 
»
Please to purchase this item. Need a web account? Register with us today!
Descripción

Descripción

gp130/CD130 Monoclonal specifically detects gp130/CD130 in Human samples. It is validated for Western Blot.
Especificaciones

Especificaciones

gp130/CD130
Western Blot 1 ug/mL
Monoclonal
Mouse
Human
P40189
3572
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human gp130 extracellular domain Leu24-Glu619 (Glu619Asp) Accession # P40189
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human gp130 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse gp130 is observed.
29104
Western Blot
Unconjugated
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CD130, CD130 antigen, CDw130, DKFZp564F053, gp130, gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form, IL-6 receptor subunit beta, IL-6R subunit beta, IL-6RB, IL-6R-beta, IL6ST, interleukin 6 signal transducer (gp130, oncostatin M receptor), interleukin receptor beta chain, interleukin-6 receptor subunit beta, Interleukin-6 signal transducer, Membrane glycoprotein 130, membrane glycoprotein gp130, Oncostatin-M receptor subunit alpha
IL6ST
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documentos

Documentos

Certificados

For Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado