Learn More
glutamate receptor, ionotropic, N-methyl D-aspartate 2C, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a full-length human GRIN2C protein.
Brand: Abnova H00002905-B01.50uL
Description
N-methyl-D-aspartate (NMDA) receptors are a class of ionotropic glutamate receptors. NMDA channel has been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDA receptor channels are heteromers composed of the key receptor subunit NMDAR1 (GRIN1) and 1 or more of the 4 NMDAR2 subunits: NMDAR2A (GRIN2A), NMDAR2B (GRIN2B), NMDAR2C (GRIN2C), and NMDAR2D (GRIN2D).
Sequence: MGGALGPALLLTSLFGAWAGLGPGQGEQGMTVAVVFSSSGPPQAQFRARLTPQSFLDLPLEIQPLTVGVNTTNPSSLLTQICGLLGAAHVHGIVFEDNVDTEAVAQILDFISSQTHVPILSISGGSAVVLTPKVHVQTHVPSCLRPGTRLGSGVLWFWEAGIRRDGQGGGGSpecifications
| glutamate receptor, ionotropic, N-methyl D-aspartate 2C | |
| Polyclonal | |
| Mouse polyclonal antibody raised against a full-length human GRIN2C protein. | |
| GRIN2C | |
| NMDAR2C/NR2C | |
| Mouse | |
| 50 μL | |
| Yes | |
| 2905 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| Unconjugated | |
| No additive | |
| BC059384 | |
| GRIN2C | |
| GRIN2C (AAH59384, 1 a.a. ∼ 171 a.a) full-length human protein. | |
| RUO | |
| Primary | |
| Human | |
| Serum |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.