missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR Mouse anti-Human, Brilliant Violet 421, Clone: G46-6, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 562804

 View more versions of this product

Product Code. 15832259

  • ONLINE EXCLUSIVE PRICE
    403.00 €
    394.83 € / Each
    Save 8.17 €
    2% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Anti++HLA-DR recognizes a human class II major histocompatibility complex (MHC) antigen. The antigen is a transmembrane glycoprotein composed of α- and β-subunits that have molecular weights of 36 and 27 kDa, respectively. Anti++HLA-DR reacts with a nonpolymorphic HLA-DR epitope and does not cross-react with HLA-DQ or HLA-DP molecules.

Host Species: Mouse
Clone: G46-6
Isotype: IgG2a, κ
Species Reactivity: Hu

Flow Cytometry

TRUSTED_SUSTAINABILITY
Specifications

Specifications

HLA-DR
Monoclonal
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Affinity Purified
RUO
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG2a κ
Flow Cytometry
G46-6
MHC class II antigen; HLA class II histocompatibility antigen
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.