missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-Human Albumin, Polyclonal, MP Biomedicals
Brand: MP Biomedicals 0855110
Code : NEW
Additional Details : Weight : 0.00200kg
Description
- Primary Antibodies
- Host: Rabbit
- Antigen: Human Albumin
- Host: rabbit
- Form: lyophilized
- Conjugate: Unlabeled
- Applications: Immunoblotting.
Specifications
Human Albumin | |
N/A | |
Rabbit | |
Primary | |
Lyophilized |
Polyclonal | |
Unconjugated | |
2 mL | |
Human |