Learn More
anti-IgG1, κ, Rat, Alexa Fluor™ 647, Clone: R3-34, BD
Brand: BD Biosciences 557865
Description
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R3-34
Isotype: IgG1 κ
Flow Cytometry, Intracellular Staining, Isotype Control
Specifications
| Flow Cytometry, Western Blot | |
| Alexa Fluor 647 | |
| Affinity Purified | |
| Undiluted at 4°C |
| R3-34 | |
| Rat | |
| 100 Tests | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.