Learn More
anti-IgG1, κ, Rat, Alexa Fluor™ 647, Clone: R3-34, BD
198.84 € - 397.67 €
Spezifikation
| Clone | R3-34 |
|---|---|
| Applications | Flow Cytometry, Western Blot |
| Conjugate | Alexa Fluor 647 |
| Host Species | Rat |
| Isotype | IgG1 κ |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
15869018
|
BD Biosciences
557731 |
0.1 mg |
414.00 € 397.67 € / Each Save 16.33 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15809088
|
BD Biosciences
557865 |
100 Tests |
207.00 € 198.84 € / Each Save 8.16 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
The immunoglobulin from the R3-34 hybridoma was identified as a non-reactive clone, following immunization with mouse Ig. The R3-34 immunoglobulin was selected as an isotype control following screening for low background on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Host Species: Rat
Clone: R3-34
Isotype: IgG1 κ
Flow Cytometry, Intracellular Staining, Isotype Control
Spezifikation
| R3-34 | |
| Alexa Fluor 647 | |
| IgG1 κ | |
| Undiluted at 4°C |
| Flow Cytometry, Western Blot | |
| Rat | |
| Affinity Purified |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.