Learn More
anti-IMPA2, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Brand:  Abnova H00003613-PW1.1Set
Product Code. 16076258
  • 835.00 € / 1 set
Includes: Antibody pair for IP: rabbit polyclonal anti-IMPA2 (300μL), Antibody pair for WB: mouse purified polyclonal anti-IMPA2 (50μg)
Specifications
IMPA2
Monoclonal/Polyclonal
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot.
IMPA2
1 Set
Primary
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-IMPA2 (300 ul) 2. Antibody pair for WB: mouse purified polyclonal anti-IMPA2 (50 ug)
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Immunoprecipitation, Western Blot
Unconjugated
IMPA2
Mouse, Rabbit
RUO
3613