missing translation for 'onlineSavingsMsg'
Learn More

Anti- Ipilimumab (Anti-Idiotype) Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18700867
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18700867 25 μg 25µg
18743487 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18700867 Supplier R&D Systems Supplier No. MAB10064SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Ipilimumab Monoclonal specifically detects Ipilimumab in Multi-Species samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ipilimumab
Applications Western Blot
Classification Monoclonal
Clone 1001345
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Host Species Mouse
Immunogen Full length Ipilimumab
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Test Specificity Detects Ipilimumab in direct ELISAs and Western blots. In Western blots, no cross-reactivity with Cetuximab, Brentuximab, Teprotumumab, and Aducanumab is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Multi-species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.