Learn More
Itch Mouse, Unlabeled, Clone: 32, BD
Mouse Monoclonal Antibody
178.69 € - 338.98 €
Specifications
| Antigen | Itch |
|---|---|
| Clone | 32 |
| Concentration | 250μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15815839
|
BD Biosciences
611198 |
50 μg |
187.00 € 178.69 € / 1 set Save 8.31 € 4% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15825839
|
BD Biosciences
611199 |
150 μg |
353.00 € 338.98 € / 1 set Save 14.02 € 3% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Maintenance of cellular function requires timely and selective degradation of key regulatory proteins. For example, progression of the mammalian cell cycle is regulated by phosphorylation/dephosphorylation and synthesis/degradation of many key proteins via the ubiquitin pathway. Ubiquitin, a soluble protein of 76 amino acids, is enzymatically attached to an ε-NH2-Lys in a target protein. Ubiquitin-conjugated proteins are recognized and degraded by the 26S proteasome. Ubiquitination requires ubiquitin-activating enzyme E1, ubiquitin-conjugating enzymes E2, and ubiquitin ligases E3. The direction of ubiquitin transfer is from E1 to E2 and from E2 to E3. Itch, a novel E3 ubiquitin ligase, is absent in the Non-agouti-lethal 18H mice. These mice develop immunological, inflammatory, epithelial, and hematopoietic diseases. Itch contains four WW protein interaction domains, which bind to proline-rich sequences in a fashion similar to SH3 domains. In addition, Itch contains a C-terminal Hect domain, which is conserved in the E3 family of ubiquitin ligases. Thus, Itch is important in the ubiquitin-dependent protein degradation occurring in normal hematopoiesis and inflammation.
Host Species: Mouse
Clone: 32
Isotype: IgG1
Species Reactivity: Rat
Immunogen: Mouse Itch aa. 114-220
Formula Weight [Chemical]: 113kDa
Immunofluorescence, Western Blotting
Specifications
| Itch | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 32 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Mouse, Rabbit, Rat | |
| Mouse Itch aa. 114-220 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.