missing translation for 'onlineSavingsMsg'
Learn More

KIT, Human, Matched Antibody Pair (AP42), Abnova™

Product Code. 16132284
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16132284 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16132284 Supplier Abnova Supplier No. H00003815AP42.1Set

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Capture antibody: mouse monoclonal anti-KIT (100μg) , Detection antibody: biotinylated mouse monoclonal anti-KIT, IgG2b Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.

To detect and quantify protein level of human

Specifications

Antigen KIT
Applications ELISA
Classification Monoclonal/Polyclonal
Conjugate Biotin
Description This antibody pair set comes with matched antibody pair to detect and quantify protein level of human KIT.
Gene KIT
Gene Alias C-Kit/CD117/PBT/SCFR
Gene Symbols KIT
Host Species Mouse
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3815
Includes Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-KIT (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-KIT, IgG2b Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Target Species Human
Content And Storage Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.