Learn More
LT-α Mouse anti-Human, Biotin, Clone: 359-81-11, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 554555
Description
The 359-81-11 antibody reacts with human Lymphotoxin-α (LT-α, also known as tumor necrosis factor-β or TNF-β). The immunogen used to generate the 359-81-11 hybridoma was recombinant human LT-α. This is a neutralizing antibody.
Host Species: Mouse
Clone: 359-81-11
Isotype: IgG1
Species Reactivity: Human
Immunogen: Recombinant Human LT-α (TNF-β)
ELISA
Specifications
| LT-α | |
| Monoclonal | |
| 0.5mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Recombinant Human LT-α (TNF-β) | |
| 0.5 mg | |
| Immunology | |
| Human | |
| IgG1 |
| ELISA | |
| 359-81-11 | |
| Biotin | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only.