missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ERK1/ERK2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen 82380
Code : A0
Additional Details : Weight : 0.00010kg
Description
It is not recommended to aliquot this antibody.
ERK1/ERK2 Polyclonal antibody specifically detects ERK1/ERK2 in Bovine, Hamster, Human, Mink, Mouse, Non-human Primate, Porcine, Rat, Yeast, Zebrafish samples. It is validated for Immunocytochemistry, Immunoprecipitation, Western Blot.Specifications
ERK1/ERK2 | |
Polyclonal | |
Unconjugated | |
MAPK3 | |
12559; 9030612K14Rik; AA407128; AU018647; BcDNA:RE08694; C78273; CG12559; CG12559-PA; CG12559-PC; CG12559-PD; CG12559-PE; CG12559-PG; CG12559-PH; CG12559-PI; CG18732; CT34260; CT39192; DERK; D-ERK; DERK-A; Diphospho-ERK; dm-dpERK; Dmel\CG12559; Dmel_CG12559; DmErk; DmERKA; DmERK-A; DmMAPK; dpERK; dp-ERK; dpERK1; dpMAPK; drosophila ERK; Dsor2; E(sina)7; EC2-1; EK2-1; enhancer of seven in absentia 7; ERK; Erk MAP kinase; Erk/Map kinase; ERK/rolled; Erk1; Erk-1; erk1 erk2; Erk1/2; ERK1b; ERK2; ERK-2; ERKA; ERK-A; ERT1; ERT2; Esrk1; extracellular signal-regulated kinase; extracellular signal-regulated kinase (ERK2); extracellular signal-regulated kinase 1; Extracellular signal-regulated kinase 2; extracellular signal-regulated kinase A; extracellular signal-regulated kinase-1; extracellular signal-regulated kinase-2; extracellular signal-related kinase 1; Extracellular-regulated kinase A; extracellular-signal-regulated kinase 2; Extracellular-signal-related kinase A; EY2-2; fi06b09; GroupI | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
100153927, 100759514, 116590, 26413, 26417, 327672, 360144, 399480, 445013, 50689, 531391, 5594, 5595 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
83 μg/mL | |
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5 | |
P46196, P27361, P28482, P63085, Q63844, P21708, P63086 | |
MAPK1, MAPK3 | |
Synthetic peptide corresponding to a sequence in the C-terminus of rat p44 MAP Kinase | |
200 μL | |
Primary | |
Bovine, Hamster, Human, Mink, Mouse, Non-human Primate, Porcine, Rat, Yeast, Zebrafish | |
Antibody | |
IgG |