missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYC-associated zinc finger protein (purine-binding transcription factor), Mouse, Clone: 6G2, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MAZ.
Brand: Abnova H00004150-M05.100ug
This item is not returnable.
View return policy
Description
Sequence: YQCPVCQQRFKRKDRMSYHVRSHDGAVHKPYNCSHCGKSFSRPDHLNSHVRQVHSTERPFKCEKCEAAFATKDRLRAHTVRHEEKVPCHVCGKMLSSAYISDHMKVHSQSpecifications
| MYC-associated zinc finger protein (purine-binding transcription factor) | |
| Monoclonal | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_002383 | |
| MAZ | |
| MAZ (NP_002374, 332 a.a. ∼ 440 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| 100 μg | |
| Transcription Regulation | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 6G2 | |
| Mouse monoclonal antibody raised against a partial recombinant MAZ. | |
| MAZ | |
| PUR1/Pur-1/SAF-1/SAF-2/ZF87/ZNF801/Zif87 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Yes | |
| 4150 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction