missing translation for 'onlineSavingsMsg'
Learn More

v-mos Moloney murine sarcoma viral oncogene homolog, Mouse, Polyclonal Antibody, Abnova™

Product Code. 16190278
Change view
Click to view available options
Quantity:
50 μL
Unit Size:
50µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16190278 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16190278 Supplier Abnova Supplier No. H00004342B01.50uL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse polyclonal antibody raised against a full-length human MOS protein.

MOS is a serine/threonine kinase that activates the MAP kinase cascade through direct phosphorylation of the MAP kinase activator MEK (MAP2K1; MIM 176872) (Prasad et al., 2008 [PubMed 18246541]).[supplied by OMIM

Sequence: MPSPLALRPYLRSEFSPSVDARPCSSPSELPAKLLLGATLPRAPRLPRRLAWCSIDWEQVCLLQRLGAGGFGSVYKATYRGVPVAIKQVNKCTKNRLASRRSFWAELNVARLRHDNIVRVVAASTRTPAGSNSLGTIIMEFGGNVTLHQVIYGAAGHPEGDAGEPHCRTGGQLSLGKCLKYSLDVVNGLLFLHSQSIVHLDLKPANILISEQDVCKISDFGCSEKLEDLLCFQTPSYPLGGTYTHRAPELLKGEGVTPKADIYSFAITLWQMTTKQAPYSGERQHILYAVVAYDLRPSLSAAVFEDSLPGQRLGDVIQRCWRPSAAQRPSARLLLVDLTSLKAELG

Specifications

Antigen v-mos Moloney murine sarcoma viral oncogene homolog
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human MOS protein.
Formulation No additive
Gene MOS
Gene Accession No. NM_005372.1
Gene Alias MGC119962/MGC119963/MSV
Gene Symbols MOS
Host Species Mouse
Immunogen MOS (NP_005363.1, 1 a.a. ∼ 346 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Cell Cycle
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 4342
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.