Learn More
anti-NCOA4, Human, Matched Antibody Pair, Abnova™
To detect and quantify protein level of human
Brand:  Abnova H00008031-AP41.1Set
Product Code. 16192814
  • 835.00 € / 1 set
Includes: Capture antibody: mouse monoclonal anti-NCOA4, IgG2b Kappa (100μg) , Detection antibody: biotinylated mouse monoclonal anti-NCOA4, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Specifications
NCOA4
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human NCOA4.
ARA70/DKFZp762E1112/ELE1/PTC3/RFG
Mouse
RUO
8031
Human
Liquid
ELISA
Biotin
NCOA4
NCOA4
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-NCOA4, IgG2b Kappa (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-NCOA4, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.