Learn More
NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7, 18kDa, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human NDUFB7 protein.
Brand: Abnova H00004713-D01P.100ug
Description
The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It is located at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. [provided by RefSeq
Sequence: MGAHLVRRYLGDASVEPDPLQMPTFPPDYGFPERKEREMVATQQEMMDAQLRLQLRDYCAHHLIRLLKCKRDSFPNFLACKQERHDWDYCEHRDYVMRMKEFERERRLLQRKKRREKKAAELAKGQGPGEVDPKVALSpecifications
| NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 7, 18kDa | |
| Polyclonal | |
| Rabbit polyclonal antibody raised against a full-length human NDUFB7 protein. | |
| NDUFB7 | |
| B18/CI-B18/MGC2480 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 4713 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with no preservative; pH 7.4 | |
| NM_004146.4 | |
| NDUFB7 | |
| NDUFB7 (NP_004137.2, 1 a.a. ∼ 137 a.a) full-length human protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.