missing translation for 'onlineSavingsMsg'
Learn More

OPA1 Mouse, Unlabeled, Clone: 18, BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 612607

 View more versions of this product

Product Code. 15826249

  • ONLINE EXCLUSIVE PRICE
    359.00 €
    344.11 € / Each
    Save 14.89 €
    4% Off

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Three major GTP-binding protein families include trimeric and low molecular weight G-proteins, as well as a family of large proteins homologous to dynamin. The dynamin family contains proteins with diverse structure and function, but highly homologous N-terminal GTPase domains. A subgroup of the dynamin G-protein-binding family includes the mitochondrial proteins Drp1/Dnm1, Mgm1, and OPA1. The latter protein is mutated in dominant optic atrophy, a disease that involves loss of visual acuity and atrophy of the optic nerve. OPA1 is expressed in heart, brain, liver, and kidney. The sequence of OPA1 includes an N-terminal region that contains a mitochondrial targeting domain and three GTP-binding motifs. The overexpression of OPA1 in Cos-7 cells shows co-localization with cytochrome c in mitochondria, and leads to alterations in mitochondrial morphology from a characteristic tubuluar shape to a vesicular pattern. Thus, OPA1 may have roles in mitochondrial biogenesis that are critical for normal cell function. This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Immunofluorescence, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

OPA1
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Human OPA1 aa. 708-830
150 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG1
Western Blot
18
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only.