missing translation for 'onlineSavingsMsg'
Learn More

P-Cadherin Mouse, Unlabeled, Clone: 56, BD

Product Code. 15805349
Change view
Click to view available options
Quantity:
150 μg
50 μg
Unit Size:
1 set
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15805349 50 μg 1 set
15815349 150 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15805349 Supplier BD Biosciences Supplier No. 610227

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Cadherins are a family of transmembrane glycoproteins involved in the Ca2+-dependent cell-cell adhesion that occurs in many tissues. Cadherins are similar in their domain structure, Ca2+ and protease sensitivity, and molecular weight. However, these proteins have distinct tissue expression patterns and immunological reactivities. P-Cadherin is localized in placenta while E-Cadherin and N-Cadherin are found in epithelial and neural tissues, respectively. P-Cadherin is an 829 amino acid polypeptide with a putative signal peptide and precursor region, an extracellular domain containing several internal repeats, and a highly hydrophobic transmembrane region. The cytoplasmic domain provides a link to the cytoskeleton through the associated catenin proteins.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen P-Cadherin
Applications Western Blot
Classification Monoclonal
Clone 56
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.